Product Details

SNP ID
rs182022575
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45595013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAACCCCGAGTTTGAGGAAACTC[G/T]CAGGGTGTTCGCCACCAAGATGGCC
Phenotype
MIM: 604543
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
LIMD1 PubMed Links

Gene Details

Gene
LIMD1
Gene Name
LIM domains containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014240.2 183 Missense Mutation CGC,CTC R45L NP_055055.1
XM_011534207.2 183 Missense Mutation CGC,CTC R45L XP_011532509.1

View Full Product Details