Product Details

SNP ID
rs182053046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63493228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGTGTCCAGGGCCTCCAGCAG[A/G]GACACGCCGCTTGCGTTGCCCTCCT
Phenotype
MIM: 602959
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EEF1A2 PubMed Links

Gene Details

Gene
EEF1A2
Gene Name
eukaryotic translation elongation factor 1 alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001958.3 1010 Silent Mutation TCC,TCT S227S NP_001949.1

View Full Product Details