Product Details

SNP ID
rs182065627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73935951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACAGCAACATTTCAGGTTACCT[A/G]GGAAAATCCTTGATTTTGGTCTCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM161B PubMed Links

Gene Details

Gene
FAM161B
Gene Name
family with sequence similarity 161 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152445.2 2191 Silent Mutation CCC,CCT P664P NP_689658.2
XM_011536475.1 2191 Silent Mutation CCC,CCT P664P XP_011534777.1
Gene
ZNF410
Gene Name
zinc finger protein 410
There are no transcripts associated with this gene.

View Full Product Details