Product Details

SNP ID
rs182088053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57436079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGACGGCGGCGACTACCGGCC[C/T]GAGCTCTCCATCCCGCTGCACCACG
Phenotype
MIM: 604894
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ONECUT2 PubMed Links

Gene Details

Gene
ONECUT2
Gene Name
one cut homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004852.2 660 Silent Mutation CCC,CCT P121P NP_004843.2
XM_017026096.1 660 Silent Mutation CCC,CCT P121P XP_016881585.1

View Full Product Details