Product Details

SNP ID
rs182093647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31083794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAATGGCCAGGTCCCCTGTCAG[C/T]AGCTGGTTGGTTGGCCTGTGGGGAA
Phenotype
MIM: 610561 MIM: 608547
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039503.2 2667 Missense Mutation TAC,TGC Y553C NP_001034592.1
XM_011545816.1 2667 Missense Mutation ACT,GCT T528A XP_011544118.1
XM_011545817.1 2667 UTR 3 XP_011544119.1
XM_011545818.2 2667 Missense Mutation TAC,TGC Y600C XP_011544120.1
XM_011545819.1 2667 Missense Mutation ACT,GCT T481A XP_011544121.1
XM_011545820.1 2667 UTR 3 XP_011544122.1
Gene
VKORC1
Gene Name
vitamin K epoxide reductase complex subunit 1
There are no transcripts associated with this gene.

Gene
ZNF646
Gene Name
zinc finger protein 646
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014699.3 2667 Intron NP_055514.3
XM_005255710.3 2667 Intron XP_005255767.1
XM_005255711.3 2667 Intron XP_005255768.1
XM_005255712.3 2667 Intron XP_005255769.1
XM_011545990.2 2667 Intron XP_011544292.1
Gene
ZNF668
Gene Name
zinc finger protein 668
There are no transcripts associated with this gene.

View Full Product Details