Product Details

SNP ID
rs181834890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:164493052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTTGTCTAGAGAGCAATCCTA[A/G]CACAATAATGTTTCAACTTGCAAGG
Phenotype
MIM: 601524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRB14 PubMed Links

Gene Details

Gene
GRB14
Gene Name
growth factor receptor bound protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303422.1 1585 Missense Mutation GCT,GTT A449V NP_001290351.1
NM_004490.2 1585 Missense Mutation GCT,GTT A536V NP_004481.2
XM_011511022.2 1585 Intron XP_011509324.1
XM_017003899.1 1585 Missense Mutation GCT,GTT A336V XP_016859388.1

View Full Product Details