Product Details

SNP ID
rs181835946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:63960720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCTCTCGGGCCAGGGCCATCT[G/T]GCGCTTCGCCTCGGCGTTGCGCTCA
Phenotype
MIM: 109760
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HTR1A PubMed Links

Gene Details

Gene
HTR1A
Gene Name
5-hydroxytryptamine receptor 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000524.3 1573 Missense Mutation AAG,CAG K334Q NP_000515.2

View Full Product Details