Product Details

SNP ID
rs181816648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51612241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTGGGCAAATCCTCACTTGCTT[A/G]TCTTGATCTCCGAGTACTCCGTGGT
Phenotype
MIM: 604200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC5 PubMed Links

Gene Details

Gene
SIGLEC5
Gene Name
sialic acid binding Ig like lectin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003830.3 1785 Missense Mutation ACA,ATA T549I NP_003821.1
XM_011527438.2 1785 UTR 3 XP_011525740.1
XM_017027419.1 1785 Missense Mutation ACA,ATA T558I XP_016882908.1

View Full Product Details