Product Details

SNP ID
rs181832842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:87518150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGACCGGCCCCTGCCCATCAGA[A/G]AGCCGTTCAGAATTTGCAGCAACAT
Phenotype
MIM: 300348
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL4 PubMed Links

Gene Details

Gene
KLHL4
Gene Name
kelch like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019117.4 439 Missense Mutation AAA,AGA K86R NP_061990.2
NM_057162.2 439 Missense Mutation AAA,AGA K86R NP_476503.1

View Full Product Details