Product Details

SNP ID
rs182426728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78971652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCCGTTGAAGTGCCCTGCCGGG[C/G]AGGGGAAGGAGGAGGTGCTCTTCGA
Phenotype
MIM: 600626
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LGALS3BP PubMed Links

Gene Details

Gene
LGALS3BP
Gene Name
galectin 3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005567.3 1861 Missense Mutation TCC,TGC S561C NP_005558.1

View Full Product Details