Product Details

SNP ID
rs182476320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149198747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCGGGCAGCAAGGGGGAGGCC[C/T]CCAAGGTATGTGGTGGTCCCTGGGG
Phenotype
MIM: 603397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF282 PubMed Links

Gene Details

Gene
ZNF282
Gene Name
zinc finger protein 282
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303481.1 708 Missense Mutation CCC,TCC P194S NP_001290410.1
NM_003575.3 708 Missense Mutation CCC,TCC P194S NP_003566.1
XM_006716151.3 708 Missense Mutation CCC,TCC P194S XP_006716214.1

View Full Product Details