Product Details

SNP ID
rs182855658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:40688349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTGGACGGCGGCGGCGACGCTT[G/T]GTGATGATGGTGATGGTGCGGGTGA
Phenotype
MIM: 608968
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAFB PubMed Links

Gene Details

Gene
MAFB
Gene Name
MAF bZIP transcription factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005461.4 892 Missense Mutation AAA,CAA K168Q NP_005452.2

View Full Product Details