Product Details

SNP ID
rs182768107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15703954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGTTTGTTTGTTTTGGTTTTT[G/T]GTTTTCTTGCCGTGGTGCAAAACTG
Phenotype
MIM: 160745 MIM: 609449
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYH11 PubMed Links

Gene Details

Gene
MYH11
Gene Name
myosin heavy chain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040113.1 6084 UTR 3 NP_001035202.1
NM_001040114.1 6084 UTR 3 NP_001035203.1
NM_002474.2 6084 UTR 3 NP_002465.1
NM_022844.2 6084 UTR 3 NP_074035.1
XM_011522502.2 6084 Intron XP_011520804.1
XM_017023250.1 6084 Intron XP_016878739.1
Gene
NDE1
Gene Name
nudE neurodevelopment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143979.1 6084 Intron NP_001137451.1
NM_017668.2 6084 Intron NP_060138.1
XM_005255396.4 6084 Intron XP_005255453.1
XM_006720897.3 6084 Intron XP_006720960.1
XM_006720900.3 6084 Intron XP_006720963.1
XM_011522553.2 6084 Intron XP_011520855.1
XM_017023349.1 6084 Intron XP_016878838.1
XM_017023350.1 6084 Intron XP_016878839.1
XM_017023351.1 6084 Intron XP_016878840.1
XM_017023352.1 6084 Intron XP_016878841.1
XM_017023353.1 6084 Intron XP_016878842.1
XM_017023354.1 6084 Intron XP_016878843.1
XM_017023355.1 6084 Intron XP_016878844.1
XM_017023356.1 6084 Intron XP_016878845.1
XM_017023357.1 6084 Intron XP_016878846.1

View Full Product Details