Product Details

SNP ID
rs182769189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241814671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTGAGGAGGGGCACGCTGGCC[C/G]GGGGCTCCGTGCGGGTGAGTGAGTG
Phenotype
MIM: 608527
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC107986004 PubMed Links

Gene Details

Gene
LOC107986004
Gene Name
uncharacterized LOC107986004
There are no transcripts associated with this gene.

Gene
NEU4
Gene Name
neuraminidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167599.2 901 Missense Mutation CGG,GGG R76G NP_001161071.1
NM_001167600.2 901 Missense Mutation CGG,GGG R63G NP_001161072.1
NM_001167601.2 901 Missense Mutation CGG,GGG R63G NP_001161073.1
NM_001167602.2 901 Missense Mutation CGG,GGG R63G NP_001161074.1
NM_080741.3 901 Missense Mutation CGG,GGG R75G NP_542779.2

View Full Product Details