Product Details

SNP ID
rs182499817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10012805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGGCATCCAGCTCCATCTCCC[A/G]TTTGGCGTCATGCTGCTCTGGGTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLEC12B PubMed Links

Gene Details

Gene
CLEC12B
Gene Name
C-type lectin domain family 12 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129998.2 336 Missense Mutation ATT,GTT I38V NP_001123470.1
NM_001319241.1 336 UTR 5 NP_001306170.1
NM_001319242.1 336 UTR 5 NP_001306171.1
NM_205852.3 336 Missense Mutation ATT,GTT I38V NP_995324.2
XM_011520658.2 336 Missense Mutation ATT,GTT I29V XP_011518960.1
XM_011520663.2 336 Missense Mutation ATT,GTT I38V XP_011518965.1
XM_017019295.1 336 UTR 5 XP_016874784.1
Gene
LOC102724020
Gene Name
uncharacterized LOC102724020
There are no transcripts associated with this gene.

View Full Product Details