Product Details

SNP ID
rs182538601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58439434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAAACTGCAGTAAGGTGGGAAG[A/C]ACAAGTAGAAAAGGCCTTCTGGCGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5B12 PubMed Links

Gene Details

Gene
OR5B12
Gene Name
olfactory receptor family 5 subfamily B member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004733.2 740 Missense Mutation GCT,TCT A240S NP_001004733.1

View Full Product Details