Product Details

SNP ID
rs182596234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:26975562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGTTGAGCGCTCTCTTGAGTA[C/T]GTTCAAAGTGATGTAGGAAAGGCTT
Phenotype
MIM: 608858 MIM: 605181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGFR1OP2 PubMed Links

Gene Details

Gene
FGFR1OP2
Gene Name
FGFR1 oncogene partner 2
There are no transcripts associated with this gene.

Gene
TM7SF3
Gene Name
transmembrane 7 superfamily member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016551.2 1733 Missense Mutation ATA,GTA I462V NP_057635.1
XM_005253391.3 1733 Missense Mutation ATA,GTA I411V XP_005253448.1
XM_011520690.2 1733 Missense Mutation ATA,GTA I340V XP_011518992.1
XM_011520691.1 1733 Intron XP_011518993.1
XM_017019463.1 1733 Missense Mutation ATA,GTA I492V XP_016874952.1
XM_017019464.1 1733 Missense Mutation ATA,GTA I360V XP_016874953.1
XM_017019465.1 1733 Missense Mutation ATA,GTA I309V XP_016874954.1
XM_017019466.1 1733 Missense Mutation ATA,GTA I289V XP_016874955.1

View Full Product Details