Product Details

SNP ID
rs182633734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128774509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGCCACTCACAGCTTTCAGGG[A/C]CCTCAGCAGCTGAGTGTAGGAGAAG
Phenotype
MIM: 603420 MIM: 613522
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALU PubMed Links

Gene Details

Gene
CALU
Gene Name
calumenin
There are no transcripts associated with this gene.

Gene
OPN1SW
Gene Name
opsin 1 (cone pigments), short-wave-sensitive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001708.2 676 Missense Mutation GCC,TCC A226S NP_001699.1

View Full Product Details