Product Details

SNP ID
rs182625553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55400775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCATTCCTAGATTTCTGATCAAC[A/T]TTGCTACAGGAGACACAACCATTTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6C65 PubMed Links

Gene Details

Gene
OR6C65
Gene Name
olfactory receptor family 6 subfamily C member 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005518.1 247 Missense Mutation ATT,TTT I83F NP_001005518.1

View Full Product Details