Product Details

SNP ID
rs182617249
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:109228399 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGTCTGTCCCAGAGAATGTGCT[A/G]AGTTTCGATGACCTTACTGCAGACG
Phenotype
MIM: 607529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SARS PubMed Links

Gene Details

Gene
SARS
Gene Name
seryl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006513.3 355 Silent Mutation CTA,CTG L85L NP_006504.2
XM_006710813.2 355 Silent Mutation CTA,CTG L85L XP_006710876.1

View Full Product Details