Product Details
- SNP ID
-
rs182388743
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:240041957 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGAAGAGGCGTAGATGAAGAGA[A/C]TTGGATGAAGAGGCATGGATGAAGG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR6B3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6732185] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR6B3
- Gene Name
- olfactory receptor family 6 subfamily B member 3
There are no transcripts associated with this gene.
- Gene
- PRR21
- Gene Name
- proline rich 21
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080835.1 |
1026 |
Missense Mutation |
AAG,AAT |
K342N |
NP_001074304.1 |
View Full Product Details