Product Details

SNP ID
rs182342705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89708491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGCTGAGGGAGGCCGCTCCTCA[G/T]CACCACGAAGGACAGGATGGGCAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPATA2L PubMed Links

Gene Details

Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
There are no transcripts associated with this gene.

Gene
VPS9D1
Gene Name
VPS9 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004913.2 3736 Silent Mutation NP_004904.2
XM_005256329.4 3736 Silent Mutation XP_005256386.1
XM_005256330.4 3736 Silent Mutation XP_005256387.1
XM_005256331.2 3736 Silent Mutation XP_005256388.1
XM_006721350.2 3736 Intron XP_006721413.1
XM_011523476.2 3736 Silent Mutation XP_011521778.1
XM_011523477.2 3736 Silent Mutation XP_011521779.1
XM_011523478.2 3736 Silent Mutation XP_011521780.1
XM_011523479.2 3736 Intron XP_011521781.1
XM_011523480.2 3736 Missense Mutation XP_011521782.1
Gene
VPS9D1-AS1
Gene Name
VPS9D1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details