Product Details

SNP ID
rs182364293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:159030723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAGAAGGAGGAACCGCAGACCC[C/T]GTAATCCCGGCTCGCGGACGGGGTC
Phenotype
MIM: 601970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00689 PubMed Links

Gene Details

Gene
LINC00689
Gene Name
long intergenic non-protein coding RNA 689
There are no transcripts associated with this gene.

Gene
VIPR2
Gene Name
vasoactive intestinal peptide receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304522.1 1477 Missense Mutation AGG,GGG R324G NP_001291451.1
NM_001308259.1 1477 Missense Mutation AGG,GGG R388G NP_001295188.1
NM_003382.4 1477 Missense Mutation AGG,GGG R404G NP_003373.2
XM_005249561.3 1477 Missense Mutation AGG,GGG R429G XP_005249618.1
XM_006716107.2 1477 Intron XP_006716170.1
XM_006716108.3 1477 Intron XP_006716171.1
XM_011516550.2 1477 Intron XP_011514852.1
XM_017012580.1 1477 Missense Mutation AGG,GGG R266G XP_016868069.1

View Full Product Details