Product Details

SNP ID
rs182365203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:54757499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCATAGTCAAGCAATTACCTTT[C/T]CTATCAGGTTTAGTGCATCTTTTAA
Phenotype
MIM: 609215
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100996654 PubMed Links

Gene Details

Gene
LOC100996654
Gene Name
uncharacterized LOC100996654
There are no transcripts associated with this gene.

Gene
SEC61G
Gene Name
Sec61 translocon gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012456.1 Intron NP_001012474.1
NM_014302.3 Intron NP_055117.1

View Full Product Details