Product Details

SNP ID
rs182373336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:53950361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTCTTATTCTTCTTCCTGGCCC[A/G]CTGTGAGTGATGCTTAGGGCTACGC
Phenotype
MIM: 112262
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMP4 PubMed Links

Gene Details

Gene
BMP4
Gene Name
bone morphogenetic protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202.4 1123 Missense Mutation CGG,TGG R300W NP_001193.2
NM_130850.3 1123 Missense Mutation CGG,TGG R300W NP_570911.2
XM_005268015.4 1123 Missense Mutation CGG,TGG R300W XP_005268072.1
XM_017021603.1 1123 Missense Mutation CGG,TGG R300W XP_016877092.1
XM_017021604.1 1123 Missense Mutation CGG,TGG R237W XP_016877093.1
XM_017021605.1 1123 Missense Mutation CGG,TGG R237W XP_016877094.1
Gene
MIR5580
Gene Name
microRNA 5580
There are no transcripts associated with this gene.

View Full Product Details