Product Details

SNP ID
rs182355867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17367279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACTGTCTACCAGTGTGTGGTAC[A/G]GCATGCGTCCTTGCATACCCCCTTG
Phenotype
MIM: 613714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCR3LG1 PubMed Links

Gene Details

Gene
NCR3LG1
Gene Name
natural killer cell cytotoxicity receptor 3 ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202439.2 2506 Missense Mutation CAG,CGG Q231R NP_001189368.1
XM_011520072.2 2506 Missense Mutation CAG,CGG Q202R XP_011518374.1
XM_011520073.2 2506 Missense Mutation CAG,CGG Q202R XP_011518375.1
XM_011520074.2 2506 Missense Mutation CAG,CGG Q202R XP_011518376.1
XM_011520075.2 2506 Missense Mutation CAG,CGG Q202R XP_011518377.1

View Full Product Details