Product Details

SNP ID
rs182394985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51480527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCACAAACCGCCCAGTGCCCAG[C/G]AGCCTGGGCCCATGTACACTGGCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM18 PubMed Links

Gene Details

Gene
CEACAM18
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278392.1 247 Missense Mutation CAG,GAG Q83E NP_001265321.1
XM_011527258.2 247 Missense Mutation CAG,GAG Q83E XP_011525560.1

View Full Product Details