Product Details

SNP ID
rs182983392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33413820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTGTCTGAGCAAGCCCCTCCT[C/T]TATGGGGACAGGTGAGACCAAGGCA
Phenotype
MIM: 616953 MIM: 603763 MIM: 602881 MIM: 603384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CUTA PubMed Links

Gene Details

Gene
CUTA
Gene Name
cutA divalent cation tolerance homolog (E. coli)
There are no transcripts associated with this gene.

Gene
KIFC1
Gene Name
kinesin family member C1
There are no transcripts associated with this gene.

Gene
PHF1
Gene Name
PHD finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002636.4 825 Silent Mutation CTC,CTT L224L NP_002627.1
NM_024165.2 825 Silent Mutation CTC,CTT L224L NP_077084.1
XM_011514662.1 825 Silent Mutation CTC,CTT L224L XP_011512964.1
XM_011514663.1 825 Silent Mutation CTC,CTT L224L XP_011512965.1
XM_011514664.1 825 Intron XP_011512966.1
XM_011514665.1 825 Silent Mutation CTC,CTT L224L XP_011512967.1
XM_011514666.1 825 Silent Mutation CTC,CTT L224L XP_011512968.1
XM_011514669.1 825 Silent Mutation CTC,CTT L224L XP_011512971.1
XM_011514670.2 825 Silent Mutation CTC,CTT L224L XP_011512972.1
XM_017010939.1 825 Intron XP_016866428.1
Gene
SYNGAP1
Gene Name
synaptic Ras GTPase activating protein 1
There are no transcripts associated with this gene.

View Full Product Details