Product Details

SNP ID
rs183236822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4947078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAAGAAGGTGGTGATTTCAACA[C/T]ATGATGTGTTGATAATGGACATGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51A2 PubMed Links

Gene Details

Gene
OR51A2
Gene Name
olfactory receptor family 51 subfamily A member 2
There are no transcripts associated with this gene.

Gene
OR51A4
Gene Name
olfactory receptor family 51 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005329.1 23 Missense Mutation TAT,TGT Y8C NP_001005329.1

View Full Product Details