Product Details

SNP ID
rs183405586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184100569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCTCTGAATTCAGTGTTTAATA[C/G]AAAGCCCTTCCGTCCGGTCACCAAC
Phenotype
MIM: 610123
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HTR3E PubMed Links

Gene Details

Gene
HTR3E
Gene Name
5-hydroxytryptamine receptor 3E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256613.1 391 Missense Mutation ACA,AGA T51R NP_001243542.1
NM_001256614.1 391 Missense Mutation ACA,AGA T66R NP_001243543.1
NM_182589.2 391 Missense Mutation ACA,AGA T66R NP_872395.2
NM_198313.2 391 Missense Mutation ACA,AGA T51R NP_938055.1
NM_198314.2 391 Missense Mutation ACA,AGA T66R NP_938056.1
Gene
HTR3E-AS1
Gene Name
HTR3E antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details