Product Details

SNP ID
rs183377548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39332851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCATTGCTAGTGTTTTACCAA[C/G]TGGCCTCTGAAGATGGTGTTCTACA
Phenotype
MIM: 601834
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCR8 PubMed Links

Gene Details

Gene
CCR8
Gene Name
C-C motif chemokine receptor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005201.3 658 Missense Mutation CTG,GTG L174V NP_005192.1

View Full Product Details