Product Details

SNP ID
rs183399390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68208266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGACCCAGAGCGCCACAAGCAA[C/G]AGGGTCAGTGCGAAGGAGGAGAAGA
Phenotype
MIM: 606725
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CALML4 PubMed Links

Gene Details

Gene
CALML4
Gene Name
calmodulin like 4
There are no transcripts associated with this gene.

Gene
CLN6
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017882.2 968 Silent Mutation CTC,CTG L270L NP_060352.1

View Full Product Details