Product Details

SNP ID
rs183332836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38802367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATGATGATGGTTCTTCGAGCT[A/G]TGAGCCCTCCTTGCAGCCTCCCGAA
Phenotype
MIM: 602518
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGALS4 PubMed Links

Gene Details

Gene
LGALS4
Gene Name
galectin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006149.3 1227 Missense Mutation ACA,ATA T203I NP_006140.1
XM_011526973.1 1227 Missense Mutation ACA,ATA T190I XP_011525275.1
XM_011526974.1 1227 Missense Mutation ACA,ATA T72I XP_011525276.1

View Full Product Details