Product Details

SNP ID
rs183084327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96131892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCTAGATCTATAATTTGACTTC[A/G]GGATATGCTTTAAATGCCCTTTAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF518A PubMed Links

Gene Details

Gene
ZNF518A
Gene Name
zinc finger protein 518A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278524.1 Intron NP_001265453.1
NM_001278525.1 Intron NP_001265454.1
NM_001278526.1 Intron NP_001265455.1
NM_014803.3 Intron NP_055618.2
XM_011540406.2 Intron XP_011538708.1
XM_011540408.2 Intron XP_011538710.1
XM_011540410.2 Intron XP_011538712.1
XM_011540412.2 Intron XP_011538714.1
XM_011540413.2 Intron XP_011538715.1
XM_011540415.2 Intron XP_011538717.1
XM_011540418.2 Intron XP_011538720.1
XM_011540419.2 Intron XP_011538721.1
XM_011540420.2 Intron XP_011538722.1
XM_017016986.1 Intron XP_016872475.1
XM_017016987.1 Intron XP_016872476.1
XM_017016988.1 Intron XP_016872477.1
XM_017016989.1 Intron XP_016872478.1
XM_017016990.1 Intron XP_016872479.1
XM_017016991.1 Intron XP_016872480.1
XM_017016992.1 Intron XP_016872481.1
XM_017016993.1 Intron XP_016872482.1
XM_017016994.1 Intron XP_016872483.1
XM_017016995.1 Intron XP_016872484.1
XM_017016996.1 Intron XP_016872485.1
XM_017016997.1 Intron XP_016872486.1
XM_017016998.1 Intron XP_016872487.1
XM_017016999.1 Intron XP_016872488.1

View Full Product Details