Product Details

SNP ID
rs183106472
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:88508598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGCTTTCCAGGGACATTCACAA[A/C]GACGAGGAGCTCTGAGTGTAATCTC
Phenotype
MIM: 604102
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GRM5 PubMed Links

Gene Details

Gene
GRM5
Gene Name
glutamate metabotropic receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000842.4 4029 Silent Mutation TCA,TCC S1179S NP_000833.1
NM_001143831.2 4029 Silent Mutation TCA,TCC S1211S NP_001137303.1
XM_006718828.3 4029 Silent Mutation TCA,TCC S1211S XP_006718891.1
XM_011542792.1 4029 Silent Mutation TCA,TCC S1211S XP_011541094.1
XM_017017627.1 4029 Silent Mutation TCA,TCC S1179S XP_016873116.1
Gene
GRM5-AS1
Gene Name
GRM5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details