Product Details

SNP ID
rs183118910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15355287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTACTGATGTCAAAGGCACTGA[A/T]ATCTCAAAGAGCTACAGCAGAGAAA
Phenotype
MIM: 300091
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FIGF PubMed Links

Gene Details

Gene
FIGF
Gene Name
c-fos induced growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004469.4 971 Silent Mutation ATA,ATT I168I NP_004460.1
Gene
PIR-FIGF
Gene Name
PIR-FIGF readthrough
There are no transcripts associated with this gene.

View Full Product Details