Product Details

SNP ID
rs183100653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9114842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAAGATCGCTAAAAAGGAAACA[A/G]CCTACCAATAAGTTTCCTCAAAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR7G1 PubMed Links

Gene Details

Gene
OR7G1
Gene Name
olfactory receptor family 7 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005192.2 922 Silent Mutation CTG,TTG L308L NP_001005192.2

View Full Product Details