Product Details

SNP ID
rs183208837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74951591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTCACACCCGCTGTATCTCAAA[C/T]AGCTTCACGTCGGTGTCGTACCAGA
Phenotype
MIM: 605752 MIM: 607828
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HID1 PubMed Links

Gene Details

Gene
HID1
Gene Name
HID1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030630.2 2490 Silent Mutation NP_085133.1
XM_005257226.1 2490 Silent Mutation XP_005257283.1
Gene
OTOP3
Gene Name
otopetrin 3
There are no transcripts associated with this gene.

View Full Product Details