Product Details

SNP ID
rs182908121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135362893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTGCTCCTCTTTGCCTAATGGC[A/G]GTCCAGTGTTGAACTCTGTAGTAAA
Phenotype
MIM: 604911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT4 PubMed Links

Gene Details

Gene
CNOT4
Gene Name
CCR4-NOT transcription complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008225.2 2465 Intron NP_001008226.1
NM_001190847.1 2465 Missense Mutation CGC,TGC R641C NP_001177776.1
NM_001190848.1 2465 Intron NP_001177777.1
NM_001190849.1 2465 Missense Mutation CGC,TGC R709C NP_001177778.1
NM_001190850.1 2465 Missense Mutation CGC,TGC R712C NP_001177779.1
NM_013316.3 2465 Missense Mutation CGC,TGC R638C NP_037448.2
XM_017012235.1 2465 Missense Mutation CGC,TGC R712C XP_016867724.1

View Full Product Details