Product Details

SNP ID
rs182944429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:106761282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGTAGATGTGCTCCTCAGCAC[A/G]CTTGGCCCTTTCAGCAGTTTTGGTT
Phenotype
MIM: 609747 MIM: 605609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABRA PubMed Links

Gene Details

Gene
ABRA
Gene Name
actin binding Rho activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139166.4 955 Missense Mutation AGT,GGT S301G NP_631905.1
Gene
OXR1
Gene Name
oxidation resistance 1
There are no transcripts associated with this gene.

View Full Product Details