Product Details

SNP ID
rs182928543
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:26180587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACACCAGGAGCTCATCCTGGGC[A/G]GGGTGGAACAGCTCCAGGCCCTGGT
Phenotype
MIM: 609121 MIM: 603272 MIM: 616698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CATSPER4 PubMed Links

Gene Details

Gene
CATSPER4
Gene Name
cation channel sperm associated 4
There are no transcripts associated with this gene.

Gene
CNKSR1
Gene Name
connector enhancer of kinase suppressor of Ras 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297647.1 245 Missense Mutation AGG,GGG R63G NP_001284576.1
NM_001297648.1 245 UTR 5 NP_001284577.1
NM_006314.2 245 Missense Mutation AGG,GGG R63G NP_006305.2
XM_011540484.2 245 UTR 5 XP_011538786.1
XM_011540485.2 245 Intron XP_011538787.1
Gene
ZNF593
Gene Name
zinc finger protein 593
There are no transcripts associated with this gene.

View Full Product Details