Product Details

SNP ID
rs183527391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35906373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGCTGCTGCTGCTGCTGCTGG[C/T]CACCTGCCTTTTCCACGGACGGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRCT1 PubMed Links

Gene Details

Gene
HRCT1
Gene Name
histidine rich carboxyl terminus 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039792.1 182 Missense Mutation GCC,GTC A29V NP_001034881.1
Gene
LINC00961
Gene Name
long intergenic non-protein coding RNA 961
There are no transcripts associated with this gene.

View Full Product Details