Product Details

SNP ID
rs183543661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:51155715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGATGGGGTCCAGCGGGGTCTT[A/G]TGGGAGAGATTATCAAGCGTTTTGA
Phenotype
MIM: 156490 MIM: 156491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NME1 PubMed Links

Gene Details

Gene
NME1
Gene Name
NME/NM23 nucleoside diphosphate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000269.2 192 Missense Mutation ATG,GTG M21V NP_000260.1
NM_198175.1 192 Missense Mutation ATG,GTG M46V NP_937818.1
Gene
NME1-NME2
Gene Name
NME1-NME2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018136.2 192 Missense Mutation ATG,GTG M21V NP_001018146.1
Gene
NME2
Gene Name
NME/NM23 nucleoside diphosphate kinase 2
There are no transcripts associated with this gene.

View Full Product Details