Product Details

SNP ID
rs183796584
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:157125614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGGGGACATTAAGGACACCTC[C/T]GCGGCCAGGGATTGGAGAGACAGCG
Phenotype
MIM: 164873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYCSP52 PubMed Links

Gene Details

Gene
CYCSP52
Gene Name
cytochrome c, somatic pseudogene 52
There are no transcripts associated with this gene.

Gene
ETV3
Gene Name
ETS variant 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145312.1 1055 Missense Mutation AGA,GGA R256G NP_001138784.1
NM_005240.1 1055 Intron NP_005231.1
XM_006711210.2 1055 Missense Mutation AGA,GGA R256G XP_006711273.1

View Full Product Details