Product Details

SNP ID
rs183866695
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:51356505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGAATTTGTCTGACTGGGTTAC[A/G]GCTTTTATAAGAAAAAAAAAAAAAG
Phenotype
MIM: 600051
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPS15 PubMed Links

Gene Details

Gene
EPS15
Gene Name
epidermal growth factor receptor pathway substrate 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159969.1 2919 UTR 3 NP_001153441.1
NM_001981.2 2919 UTR 3 NP_001972.1
XM_005270618.2 2919 UTR 3 XP_005270675.1
XM_017000615.1 2919 UTR 3 XP_016856104.1
XM_017000616.1 2919 UTR 3 XP_016856105.1
XM_017000617.1 2919 UTR 3 XP_016856106.1
XM_017000618.1 2919 UTR 3 XP_016856107.1
XM_017000619.1 2919 UTR 3 XP_016856108.1
XM_017000620.1 2919 UTR 3 XP_016856109.1

View Full Product Details