Product Details

SNP ID
rs183631659
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:247256082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGATCATAGAGATGTTGAAAT[C/T]GGTTATTATTGAGAACATGGCAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VN1R5 PubMed Links

Gene Details

Gene
VN1R5
Gene Name
vomeronasal 1 receptor 5 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173858.1 11 Missense Mutation TCG,TTG S4L NP_776257.1

View Full Product Details