Product Details

SNP ID
rs183607155
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:13475996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAATGATGATGCAGAGGTCTGT[A/G]AGTGACAGGCTGAACCTACACTCGC
Phenotype
MIM: 615212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC38 PubMed Links

Gene Details

Gene
LRRC38
Gene Name
leucine rich repeat containing 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010847.1 889 Silent Mutation CTC,CTT L245L NP_001010847.1

View Full Product Details