Product Details

SNP ID
rs183776114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:98875418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTATATATTGTAACCTTACCTA[A/G]TATGTCCAGCTGCTGTAACCTGGTA
Phenotype
MIM: 605654
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL4 PubMed Links

Gene Details

Gene
FBXL4
Gene Name
F-box and leucine rich repeat protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278716.1 2124 Silent Mutation CTA,TTA L567L NP_001265645.1
NM_012160.4 2124 Silent Mutation CTA,TTA L567L NP_036292.2
XM_005266930.2 2124 Silent Mutation CTA,TTA L543L XP_005266987.1
XM_011535748.2 2124 Intron XP_011534050.1
XM_017010726.1 2124 Silent Mutation CTA,TTA L567L XP_016866215.1
XM_017010727.1 2124 Silent Mutation CTA,TTA L543L XP_016866216.1
XM_017010728.1 2124 Silent Mutation CTA,TTA L325L XP_016866217.1

View Full Product Details