Product Details

SNP ID
rs183773199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9186354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCCTCCAGCTGGCCTGCTCTGA[C/T]ACCTTCCTGAACAGCACGTTGATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7D2 PubMed Links

Gene Details

Gene
OR7D2
Gene Name
olfactory receptor family 7 subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175883.2 761 Silent Mutation GAC,GAT D191D NP_787079.1

View Full Product Details