Product Details

SNP ID
rs183472636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40126326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTGAGAAAATTAAACTGGAGTG[C/T]CAGCCGGAGCTTTCCGAGACATCCC
Phenotype
MIM: 614801
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSL1 PubMed Links

Gene Details

Gene
MSL1
Gene Name
male specific lethal 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012241.1 464 Silent Mutation TGC,TGT C41C NP_001012241.1
XM_005257298.4 464 Silent Mutation TGC,TGT C304C XP_005257355.1
XM_005257299.4 464 Silent Mutation TGC,TGT C304C XP_005257356.3

View Full Product Details